missing translation for 'onlineSavingsMsg'
Learn More
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals Product Code.: 10666184

(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals

Product Code. 10666184
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10666184

Brand: Thermo Scientific Acros 295770050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
Molecular Weight (g/mol) 128
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chlorideShow MoreShow Less
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride
SMILES C[N+](=CCl)C.[Cl-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 139°C
Color Brown to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCH=N(CH3)2Cl
Quantity 5 g
Fieser 04,186; 05,251; 06,220; 07,422; 08,186; 09,514; 10,457; 12,564; 13,341
Solubility Information Solubility in water: reacts. Other solubilities: 28g/100mL chloroform,slightly soluble in other solvents
Formula Weight 128
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material (Chloromethylene)dimethylammonium chloride, 96%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442060 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number295770250,295770050
Lot NumberA0428808 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog Number295770250,295770050
Lot NumberA0422498 Certificate TypeCertificate of Analysis Date15/10/2020 Catalog Number295770250,295770050
Lot NumberA0385409 Certificate TypeCertificate of Analysis Date10/06/2020 Catalog Number295770250,295770050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Chloromethylene)dimethylammonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.