missing translation for 'onlineSavingsMsg'
Learn More
Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals Product Code.: 11315978

Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals

Product Code. 11315978
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11315978

Brand: Thermo Scientific Alfa Aesar 010468.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chlorotris(triphenylphosphine)rhodium(I) is used as a homogeneous hydrogenation catalyst. It is used in the selective hydrogenation of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14694-95-2
Molecular Formula C54H45ClP3Rh
Molecular Weight (g/mol) 925.23
MDL Number MFCD00010016
InChI Key IXAYKDDZKIZSPV-UHFFFAOYSA-M
Synonym chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, rhodium; triphenylphosphane; chlorideShow MoreShow Less
PubChem CID 84599
IUPAC Name rhodium;triphenylphosphane;chloride
SMILES [Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1Show MoreShow Less

Specifications

Melting Point ∼250°C (decomposition)
Color Red
Odor Odorless
Linear Formula RhCl[(C6H5)3P]3
Quantity 5 g
Merck Index 14,10047
Solubility Information Soluble in most solvents (e;g; benzene,ethanol,chloroform,dichloromethane) but with phosphine dissociation; Reacts with O2 in solution
Formula Weight 925.23
Percent Purity 97%
Physical Form Micro Crystals
Chemical Name or Material Chlorotris(triphenylphosphine)rhodium(I)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE21I06 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010482.03,010468.03,010468.06
Lot NumberE29T022 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010482.03,010468.03,010468.06
Lot NumberY19B046 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010482.03,010468.03,010468.06
Lot NumberI05H08 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010482.03,010468.03,010468.06
Lot Number10194199 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010482.03,010468.03,010468.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.