missing translation for 'onlineSavingsMsg'
Learn More
Chromium hexacarbonyl, 99+%, Thermo Scientific Chemicals Product Code.: 10687362

Chromium hexacarbonyl, 99+%, Thermo Scientific Chemicals

Product Code. 10687362
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10687362

Brand: Thermo Scientific Acros 221050100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-92-6
Molecular Formula C6CrO6
Molecular Weight (g/mol) 220.06
MDL Number MFCD00010945
InChI Key KOTQLLUQLXWWDK-UHFFFAOYSA-N
Synonym chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10gShow MoreShow Less
PubChem CID 25589
ChEBI CHEBI:33031
IUPAC Name carbon monoxide;chromium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Boiling Point 220°C
Melting Point 150°C
Color White through Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 10 g
Assay Percent Range 99+%
Linear Formula Cr(CO)6
Merck Index 15, 2236
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in organic solvents
Formula Weight 220.06
Percent Purity ≥99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Chromium hexacarbonyl
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466774 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number221050100,221050500
Lot NumberA0461455 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number221050100,221050500
Lot NumberA0455679 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number221050100,221050500
Lot NumberA0449247 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number221050100,221050500
Lot NumberA0408405 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number221050100,221050500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chromium hexacarbonyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H350i-May cause cancer by inhalation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Chromium carbonyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium hexacarbonyl, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.