missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose
The ChromoTek GFP-Trap™ Magnetic Agarose are affinity beads for IP of GFP-fusion proteins.
£208.00 - £5160.00
Specifications
Binding Capacity | 10 μL slurry bind more than 8 μg of recombinant GFP |
---|---|
Content And Storage | Upon receipt store at 4°C. Do not freeze. |
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP |
Label or Dye | Magnetic Agarose |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17313363
|
Proteintech
gtma-10 |
10 Reactions |
£208.00
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17323363
|
Proteintech
GTMA-100 |
100 Reactions |
£1572.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17333363
|
Proteintech
gtma-20 |
20 Reactions |
£361.92
20 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17343363
|
Proteintech
gtma-200 |
200 Reactions |
£2828.00
200 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17353363
|
Proteintech
gtma-400 |
400 Reactions |
£5160.00
400 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
10 μL slurry bind more than 8 μg of recombinant GFP | |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose |
Upon receipt store at 4°C. Do not freeze. | |
AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP | |
Alpaca |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose