Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose
The ChromoTek GFP-Trap™ Magnetic Agarose are affinity beads for IP of GFP-fusion proteins.
Brand: Proteintech gtma-10
Description
- Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
10 μL slurry bind more than 8 μg of recombinant GFP | |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
10 Reactions |
Upon receipt store at 4°C. Do not freeze. | |
AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP | |
Alpaca |
Product Suggestions
Customers who viewed this item also viewed.