missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 20mm Crimp Cap

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™ 20 02 2515

 View more versions of this product

Product Code. 15373696

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • 20mm Septa
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Septa
20 mm
1000
Septa
White
PTFE Virginal
0.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Septum for 20mm Crimp Cap > PTFE virginal white,0.2 mm thickness,53° shore D

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.