Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose
The ChromoTek Halo-Trap Magnetic Agarose consists of an anti-Halo-tag Nanobody (VHH), which is covalently bound to magnetic agarose beads.
Brand: Proteintech OTMA-100
Additional Details : Weight : 0.00120kg
Description
- Halo-Trap Magnetic Agarose is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand
- The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
12.5 μg of recombinant Halo-tag per 25 μL bead slurry | |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
100 Reactions |
Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature. | |
Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands | |
Alpaca |