missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose
The ChromoTek Halo-Trap Magnetic Agarose consists of an anti-Halo-tag Nanobody (VHH), which is covalently bound to magnetic agarose beads.
£208.00 - £5160.00
Specifications
Binding Capacity | 12.5 μg of recombinant Halo-tag per 25 μL bead slurry |
---|---|
Content And Storage | Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature. |
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands |
Label or Dye | Magnetic Agarose |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303413
|
Proteintech
OTMA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17313413
|
Proteintech
OTMA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17323413
|
Proteintech
OTMA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17333413
|
Proteintech
OTMA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17343413
|
Proteintech
OTMA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
- Halo-Trap Magnetic Agarose is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand
- The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
12.5 μg of recombinant Halo-tag per 25 μL bead slurry | |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose |
Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature. | |
Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands | |
Alpaca |