Learn More
Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose
The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins.
Brand:  Proteintech NTMA-400
Additional Details : Weight : 0.00480kg
 View more versions of this product
Product Code. 17313403
  • £5160.00 / 400 reactions
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mNeonGreen, Branchiostoma lanceolatum
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
400 Reactions