Learn More
Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose
The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins.
Brand: Proteintech NTMA-10
Additional Details : Weight : 0.00240kg
Description
- Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mNeonGreen, Branchiostoma lanceolatum | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
10 Reactions |