Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Agarose
The ChromoTek RFP-Trap™ Magnetic Agarose are affinity beads for IP of RFP-fusion proteins.
Brand: Proteintech rtma-200
This item is not returnable.
View return policy
Description
- Comprises a RFP Nanobody/ VHH coupled to magnetic agarose
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
200 Reactions |