missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Agarose
The ChromoTek RFP-Trap™ Magnetic Agarose are affinity beads for IP of RFP-fusion proteins.
£208.00 - £5160.00
Specifications
Content And Storage | Upon receipt store at 4°C. Do not freeze. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet |
Label or Dye | Magnetic Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17343453
|
Proteintech
RTMA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17353453
|
Proteintech
RTMA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17363453
|
Proteintech
RTMA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
17373453
|
Proteintech
RTMA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17383453
|
Proteintech
RTMA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
- Comprises a RFP Nanobody/ VHH coupled to magnetic agarose
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose |