Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose
Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins.
Brand: Proteintech ETMA-400
Additional Details : Weight : 0.00480kg
Description
- Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
Spot-Tag Sequence Motif PDRVRAVSHWSS | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
400 Reactions |