missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose
Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins.
£208.00 - £5160.00
Specifications
Content And Storage | Upon receipt store at 4°C. Do not freeze. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | Spot-Tag Sequence Motif PDRVRAVSHWSS |
Label or Dye | Magnetic Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303323
|
Proteintech
ETMA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17313323
|
Proteintech
ETMA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17373313
|
Proteintech
ETMA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17383313
|
Proteintech
ETMA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17393313
|
Proteintech
ETMA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
- Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
Spot-Tag Sequence Motif PDRVRAVSHWSS | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose |