Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose
Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins.
Brand:  Proteintech ETMA-10
Additional Details : Weight : 0.00240kg
 View more versions of this product
Product Code. 17373313
  • £208.00 / 10 reactions
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
Spot-Tag Sequence Motif PDRVRAVSHWSS
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
10 Reactions