Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose
TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.
Brand:  Proteintech TBTMA-100
Additional Details : Weight : 0.00120kg
 View more versions of this product
Product Code. 17313523
  • £1572.00 / 100 reactions
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green
Alpaca
IP, CoIP, ELISA
Magnetic Agarose
100 Reactions