missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0 Product Code.: 15433999

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0

Product Code. 15433999
500 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15433999

Brand: Thermo Scientific Chemicals J61391.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citrate is an intermediate in the TCA cycle and fatty acid synthesis. It is an allosteric modulator of acetyl-CoA carboxylase, the enzyme that regulates the conversion of acetyl-CoA to malonyl-CoA. It has anticoagulant activity and is a calcium chelator. Its function is to break protein cross-links; thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies.

Solubility
Fully miscible in water. Insoluble in ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6132-04-3

Specifications

Color Colorless
Concentration 0.5M buffer soln.
Molecular Weight (g/mol) 294.10
pH 3.0
Quantity 500 mL
Odor Odorless
Content And Storage Keep cold
Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX29K537 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ61391.AK,J61391.AP
Lot NumberU25K503 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberJ61391.AK,J61391.AP
Lot NumberQ29K517 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ61391.AK,J61391.AP
Lot NumberP24K524 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ61391.AK,J61391.AP
Lot NumberP02K554 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberJ61391.AK,J61391.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 3.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.