missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid, trisodium salt 98%, anhydrous, pure, Thermo Scientific Chemicals
£19.60 - £154.00
Chemical Identifiers
CAS | 68-04-2 |
---|---|
Molecular Formula | C6H5Na3O7 |
Molecular Weight (g/mol) | 258.07 |
MDL Number | MFCD00012462 |
InChI Key | HRXKRNGNAMMEHJ-UHFFFAOYSA-K |
Synonym | sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous |
PubChem CID | 6224 |
ChEBI | CHEBI:53258 |
IUPAC Name | trisodium;2-hydroxypropane-1,2,3-tricarboxylate |
SMILES | [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10362234
|
Thermo Scientific
391971000 |
100 g | Plastic bottle |
£19.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360334
|
Thermo Scientific Acros
391975000 |
500 g | Plastic bottle |
£48.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10045303
|
Thermo Scientific Acros
391970025 |
2.5 kg | Plastic bottle |
£154.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68-04-2 | |
258.07 | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
6224 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate |
C6H5Na3O7 | |
MFCD00012462 | |
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous | |
CHEBI:53258 | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
Specifications
68-04-2 | |
White | |
Authentic | |
C6H5Na3O7 | |
15, 8737 | |
Solubility in water: 500g/L (20°C). | |
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
258.07 | |
CHEBI:53258 | |
0.98 | |
Crystalline Powder |
300.0°C | |
290°C | |
0.98 | |
MFCD00012462 | |
sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrous | |
HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate | |
6224 | |
258.07 | |
Pure | |
Citric acid, trisodium salt |
RUO – Research Use Only