missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS

Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes, Class AS come with blue graduation.

Brand:  Hirschmann™ 1341140

 View more versions of this product

Product Code. 11780784

  • £115.00 / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Acc. to DIN EN ISO 648
  • One mark, blue graduation
  • Conformity certified, with dated batch identification
Specifications

Specifications

AS
Yes
6/Pk.
2 mL
Volumetric Pipet
AR-GLAS
Blue
0.01 mL
350 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS > Capacity: 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.