Learn More
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR™ glass.
Brand:  Hirschmann™ 1340175
 View more versions of this product
Product Code. 11738001
  • £141.00 / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One mark
  • Acc. to DIN EN ISO 648
  • Conformity certified, with dated batch identification
Specifications
Specifications
AS
Yes
6/Pk.
50 mL
Volumetric Pipet
AR-GLAS
Brown
0.03 mL
550 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation > Capacity: 50mL