Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
Brand: Hirschmann™ 1300150
Description
- One mark
- Amber stain graduation
- Acc. to DIN EN ISO 648
Specifications
B | |
Yes | |
6/Pk. | |
4 mL | |
Volumetric Pipet |
Glass | |
Brown | |
0.023 mL | |
410 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.