missing translation for 'onlineSavingsMsg'
Learn More
Purus™ Pure Comfort Cleanroom Socks Product Code.: 17793799
Estimated Shipment: 06-05-2025
to see stock.
Add to basket

Purus™ Pure Comfort Cleanroom Socks

Product Code. 17793799 Shop All Purus Products
Reusable, Small
Click to view available options
Disposable:
Reusable
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Product Code. 17793799

Brand: Purus™ CRSLS

Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Purus™ Cleanroom Socks are designed for cleanroom use and comfort.

  • Available in both disposable and launderable styles
  • Made from 100% polyester
  • Cleanroom compatible
  • Comfortable for extended use
  • No particle generation

Specifications

Color White
For Use With (Application) Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors
Material Polyester
Size Small
Disposable Reusable
Sterility Non-sterile
Packaging Quantity 1 Pair/Bag, 200 Pair/Cs
Product Type Launderable Cleanroom Socks
Cleanroom Class Class 10-1000 (ISO 4-6)
Packaging Type 200 Pair/Cs

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3005 Certificate TypeCertificate of Compliance Date15/01/2025 Catalog NumberCRS-L-S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Purus™ Pure Comfort Cleanroom Socks >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.