Learn More
Cytiva CNBr-Activated Sepharose™ Medium
Use for coupling antibodies or other large proteins containing -NH2 groups to the Sepharose media. CNBr-Activated Sepharose™ Medium is a pre-activated media perfect for custom affinity applications.
Brand:  Cytiva 17-0430-01
Product Code. 11514905
  • £405.00 / 15g
Explore more special offers
Description
Description
  • Allows spontaneous and rapid coupling of proteins without the need for an intermediate spacer arm
  • Multipoint attachment resulting in a chemically stable product
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large Protein
90 μm
15 g
For coupling ligands containing primary amino groups
4% Agarose
2 to 11
2°C to 8°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva CNBr-Activated Sepharose™ Medium > 15g