Learn More
Thermo Scientific™ Remel™ Candida albicans Test Kit
Thermo Scientific™ Remel Candida albicans Test Kit is a reagent impregnated disc test for presumptive identification of Candida albicans.
Brand: Thermo Scientific™ R30851401
212.46 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Remel Candida albicans Test Kit is a reagent impregnated disc test used for rapid presumptive identification of Candida albicans.
- Rapid test for the detection of enzymes L-proline aminopeptidase and Β-galactosaminidase in yeast cells for the presumptive identification of Candida albicans cultured on laboratory media
A rapid and sensitive method utilizing a combination of p-nitrophenyl-N-acetyl-Β-D-galactosaminide (NGL) and L-proline-Β-naphthylamide (PRO)
Includes:
50 test discs; 1 × 45mL vial 03% sodium hydroxide; 1 vial color developer; 50 plastic tubes with caps; wooden applicator sticks.

Specifications
Unopened reagents stable until the expiration date when stored at 2°C to 8°C | |
Candida albicans Test Kit | |
50 Tests |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6184873 | Certificate of Analysis | 07/02/2025 | R30851401 |
6142100 | Certificate of Analysis | 19/11/2024 | R30851401 |
3811519 | Certificate of Analysis | 20/08/2024 | R30851401 |
3786644 | Certificate of Analysis | 22/05/2024 | R30851401 |
3769928 | Certificate of Analysis | 05/04/2024 | R30851401 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For in vitro diagnostic use