missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiCard™ Candida Kit

Thermo Scientific™ Remel BactiCard Candida Kit is used for presumptive identification of Candida albicans using enzyme technology.

Brand:  Thermo Scientific™ R21106

215.43 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 13235659

  • £384.00 / 25 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Remel BactiCard Candida Kit is used for presumptive identification of Candida albicans using enzyme technology.

  • For use in qualitative procedures for the presumptive identification of Candida albicans by the rapid detection of L-Proline aminopeptidase and Β-galactosaminidase
  • Test results in 30 seconds
  • 4-Methylumbelliferyl-N-acetyl-Β-D-galactosaminide in the presence of the enzyme Β-galactosaminidase releases 4-methylumbelliferone, which is a highly fluorescent compound detectable by ultraviolet light
  • L-Proline aminopeptidase hydrolyzes L-Proline-Β-napthylamide and reacts with the color developer to form a red color If PRO and MUGAL are both positive, then C. albicans is present Product is ready for use and no further preparation is necessary

Includes:

Test cards, rehydrating fluid, color developer, MUGAL reagent and applicator sticks.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in its original container at 2° to 8°C until used.
BactiCard™ Candida
25 Tests/Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6163818 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberR21106
Lot Number6127425 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberR21106
Lot Number3742023 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog NumberR21106
Lot Number3656307 Certificate TypeCertificate of Analysis Date17/08/2023 Catalog NumberR21106
Lot Number3575450 Certificate TypeCertificate of Analysis Date20/01/2023 Catalog NumberR21106
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Remel™ BactiCard™ Candida Kit > 25 Tests/Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For in vitro diagnostic use