missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution

For routine hemostasis testing

Brand:  Greiner Bio-One 454322


 View more versions of this product

Product Code. 13078692

  • £229.99 / Pack of 1200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

 

  • 13 × 75mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium Citrate Blood Collection Tube
PET
Blue with White Ring
3.2% Citrate Solution
1.8 mL
13 x 75 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Blood tube Vacuette 3.2% buffered citrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Citric Acid Monohydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution > 3.2% Citrate Solution; Fill volume: 1.8mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.