missing translation for 'onlineSavingsMsg'
Learn More
Cobalt aluminum oxide, Co 39-42%, Thermo Scientific Chemicals Product Code.: 11363387

Cobalt aluminum oxide, Co 39-42%, Thermo Scientific Chemicals

Product Code. 11363387
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11363387

Brand: Thermo Scientific Alfa Aesar 017686.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the catalytic deposition of nitrous oxide over calcinated cobalt aluminum hydrotalcites. The activity increased with an increase in cobalt concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12672-27-4
Molecular Formula Al2Co2O5
Molecular Weight (g/mol) 251.824
MDL Number MFCD00016016
InChI Key CRHLEZORXKQUEI-UHFFFAOYSA-N
Synonym cobalt aluminate, aluminum cobalt oxide, aluminum cobalt oxide al2coo4, cobalt aluminate 100g, dialuminium cobalt tetraoxide, dialuminum; cobalt 2+ ; oxygen 2-, dialuminium 3+ dicobalt 2+ pentakis oxide 2-, dialuminium 3+ ion bis ? 2-cobalt 2+ ion pentaoxidandiideShow MoreShow Less
PubChem CID 159384
IUPAC Name dialuminum;cobalt(2+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[O-2].[O-2].[Al+3].[Al+3].[Co+2].[Co+2]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 176.89
Concentration or Composition (by Analyte or Components) Co 39 to 41%
Odor Odorless
Chemical Name or Material Cobalt aluminum oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR01K001 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number017686.22,017686.36
Lot NumberS21I048 Certificate TypeCertificate of Analysis Date01/07/2022 Catalog Number017686.22,017686.36
Lot NumberP14I055 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number017686.22,017686.36
Lot NumberP08I002 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number017686.22,017686.36
Lot NumberB05J32 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number017686.22,017686.36
compliance-icons compliance-icons
Product Identifier
  • Cobalt aluminum oxide
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt aluminum oxide, Co 39-42%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.