Learn More
Cobalt Chromium Iron Nickel Molybdenum Manganese wire, 0.25mm (0.01 in.) dia., Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 045783.G6
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Cobalt Chromium Iron Nickel Molybdenum Manganese wire | |
1450°C to 1460°C | |
MFCD18785637 | |
Insoluble in water. | |
Wire |
0.25mm (0.01 in.) dia., Hard | |
Co:Cr:Fe:Ni:Mo:Mn; 40:20:15:15:7:2 wt% | |
10 m | |
≈8.1g/5m |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Cobalt Chromium Iron Nickel Molybdenum Manganese wire
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Cobalt, Nickel
Your input is important to us. Please complete this form to provide feedback related to the content on this product.