Learn More
Cobalt(II) carbonate, 99% (metals basis), Co 45% min, Thermo Scientific Chemicals
For Use in Ceramics, Manufacturing Co Compounds, Pigments
£57.66 - £134.67
Chemical Identifiers
CAS | 513-79-1 |
---|---|
Molecular Formula | CCoO3 |
Molecular Weight (g/mol) | 118.94 |
MDL Number | MFCD00010940 |
InChI Key | ZOTKGJBKKKVBJZ-UHFFFAOYSA-L |
Synonym | cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuvShow More |
PubChem CID | 10565 |
IUPAC Name | cobalt(2+);carbonate |
SMILES | [Co++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391109
|
Thermo Scientific Alfa Aesar
011343.22 |
100 g |
£57.66
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381109
|
Thermo Scientific Alfa Aesar
011343.36 |
500 g |
£134.67
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-79-1 | |
118.94 | |
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L | |
10565 | |
[Co++].[O-]C([O-])=O |
CCoO3 | |
MFCD00010940 | |
cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuvShow More | |
cobalt(2+);carbonate |
Specifications
(decomposition) | |
Pink | |
99% (Metals basis, Co 45% min) | |
MFCD00010940 | |
Insoluble in water,alcohol and methyl acetate. | |
[Co++].[O-]C([O-])=O | |
118.94 | |
118.94 | |
4.13 |
513-79-1 | |
Powder | |
CCoO3 | |
cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuv | |
ZOTKGJBKKKVBJZ-UHFFFAOYSA-L | |
cobalt(2+);carbonate | |
10565 | |
Odorless | |
Cobalt(II) carbonate |
Safety and Handling
P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H341-H350i-H360F
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 208-169-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cobalt(II) carbonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H360F-May damage fertility.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Carbonic acid, cobalt(2+) salt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.