Learn More
Cobalt(II) carbonate, 99% (metals basis), Co 45% min, Thermo Scientific Chemicals
Description
Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | Pink |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99% (Metals basis, Co 45% min) |
Solubility Information | Insoluble in water,alcohol and methyl acetate. |
Formula Weight | 118.94 |
Odor | Odorless |
Density | 4.13 |
Chemical Name or Material | Cobalt(II) carbonate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z23I006 | Certificate of Analysis | 06/01/2023 | 011343.36,011343.22 |
Z23I005 | Certificate of Analysis | 06/01/2023 | 011343.36,011343.22 |
Y03A020 | Certificate of Analysis | 12/11/2021 | 011343.36,011343.22 |
E10E03 | Certificate of Analysis | 12/11/2021 | 011343.36,011343.22 |
P26E038 | Certificate of Analysis | 12/11/2021 | 011343.36,011343.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.