missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) carbonate hydrate, Thermo Scientific Chemicals Product Code.: 11994311

Cobalt(II) carbonate hydrate, Thermo Scientific Chemicals

Product Code. 11994311
1 kg, Plastic bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11994311

Brand: Thermo Scientific Acros 192100010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57454-67-8
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00149654
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
PubChem CID 16211458
SMILES [Co++].[O-]C([O-])=O

Specifications

Color White to Yellow
Physical Form Mass, Crystals, Crystalline Powder and/or Chunks
Quantity 1 kg
Linear Formula CoCO3·xH2O
Merck Index 14,2436
Solubility Information Solubility in water: insoluble
IUPAC Name cobalt(2+);carbonate;hydrate
Formula Weight 118.94
Percent Purity ≥45.5% (Cobalt) (Complexometry)
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) carbonate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461099 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number192102500,192100500,192100010
Lot NumberA0451066 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number192102500,192100500,192100010
Lot NumberA0392923 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number192102500,192100500,192100010
Lot NumberA0406277 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number192102500,192100500,192100010
Lot NumberA0411231 Certificate TypeCertificate of Analysis Date11/02/2022 Catalog Number192102500,192100500,192100010
compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) carbonate hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) carbonate hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.