missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(Ii) Chloride Hexahydrate, puriss. p.a. ACS Reagent, Honeywell™ Product Code.: 15673370

Cobalt(Ii) Chloride Hexahydrate, puriss. p.a. ACS Reagent, Honeywell™

Product Code. 15673370 Shop All Honeywell Fluka Products
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 15673370

Brand: Honeywell-Fluka 31277100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reagent European Pharmacopoeia, 98-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7791-13-1
Quantity 100 g
Molecular Formula Cl2CoH12O6
Linear Formula CoCl2 · 6H2O
MDL Number MFCD00149652
UN Number 3077
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
Molecular Weight (g/mol) 237.92
Formula Weight 237.93g/mol
Percent Purity 98 to 102%
Grade ACS/puriss. p.a.
Vapor Pressure 40mmHg (0°C)
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cobalt dichloride hexahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(Ii) Chloride Hexahydrate, puriss. p.a. ACS Reagent, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.