Learn More
Cobalt(II) chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1049°C |
Melting Point | 735°C |
Color | Blue |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.998% (Metals basis) |
Solubility Information | Freely soluble in water and methanol. Soluble in alcohol,acetone,ether,glycerol,and pyridine. |
Formula Weight | 129.84 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P22K011 | Certificate of Analysis | 08/04/2024 | 013665.06,013665.03,013665.14 |
Q21D126 | Certificate of Analysis | 12/04/2022 | 013665.06,013665.03,013665.14 |
E16F21 | Certificate of Analysis | 08/12/2021 | 013665.06,013665.03,013665.14 |
U16H091 | Certificate of Analysis | 08/12/2021 | 013665.06,013665.03,013665.14 |
K09J19 | Certificate of Analysis | 07/12/2021 | 013665.06,013665.03,013665.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.