Learn More
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
£56.30 - £653.00
Chemical Identifiers
CAS | 1308-06-1 |
---|---|
Molecular Formula | Co3O4 |
Molecular Weight (g/mol) | 240.80 |
MDL Number | MFCD00010939 |
InChI Key | UBEWDCMIDFGDOO-UHFFFAOYSA-N |
Synonym | oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt |
PubChem CID | 11651651 |
IUPAC Name | oxocobalt;oxo(oxocobaltiooxy)cobalt |
SMILES | [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419850
|
Thermo Scientific Alfa Aesar
044661.14 |
25 g |
£56.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429850
|
Thermo Scientific Alfa Aesar
044661.22 |
100 g |
£156.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439850
|
Thermo Scientific Alfa Aesar
044661.36 |
500 g |
£653.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-06-1 | |
240.80 | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
11651651 | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Co3O4 | |
MFCD00010939 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
oxocobalt;oxo(oxocobaltiooxy)cobalt |
Specifications
900°C (decomposition) | |
Black | |
Co3O4 | |
UN3077 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
oxocobalt;oxo(oxocobaltiooxy)cobalt | |
11651651 | |
99% | |
(metals basis) |
1308-06-1 | |
Powder | |
MFCD00010939 | |
14,2429 | |
Soluble in acids and alkali. Insoluble in water. | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] | |
240.80 | |
240.8 | |
6.11 g/mL | |
Cobalt (II,III) oxide |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H350i
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 215-157-2
missing translation for 'rtecsNumber' : GG2500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only