Learn More
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals
Description
Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 900°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN3077 |
Merck Index | 14,2429 |
Solubility Information | Soluble in acids and alkali. Insoluble in water. |
Formula Weight | 240.8 |
Percent Purity | 99% |
Density | 6.11 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y15K027 | Certificate of Analysis | 19/11/2024 | 044661.22,044661.14,044661.36 |
X14K050 | Certificate of Analysis | 05/11/2024 | 044661.22,044661.14,044661.36 |
U30K017 | Certificate of Analysis | 18/09/2024 | 044661.22,044661.14,044661.36 |
Q23K072 | Certificate of Analysis | 11/06/2024 | 044661.22,044661.14,044661.36 |
W01H016 | Certificate of Analysis | 22/11/2021 | 044661.22,044661.14,044661.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.