missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11439850

Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11439850
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11439850

Brand: Thermo Scientific Alfa Aesar 044661.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobaltShow MoreShow Less
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 500 g
UN Number UN3077
Merck Index 14,2429
Solubility Information Soluble in acids and alkali. Insoluble in water.
Formula Weight 240.8
Percent Purity 99%
Density 6.11 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt (II,III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY15K027 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number044661.22,044661.14,044661.36
Lot NumberX14K050 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number044661.22,044661.14,044661.36
Lot NumberU30K017 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number044661.22,044661.14,044661.36
Lot NumberQ23K072 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number044661.22,044661.14,044661.36
Lot NumberW01H016 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044661.22,044661.14,044661.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II,III) oxide, nanopowder, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.