missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals Product Code.: 11397878

Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Product Code. 11397878
500 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11397878

Brand: Thermo Scientific Alfa Aesar 036418.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Battery Additive, in the Manufacture of Cobalt Pigments and Invisible Inks, For Decorating Stoneware and Porcelain

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications:
  • As battery additive, in manufacture of cobalt pigments and invisible inks, for decorating stoneware and procelain, and in preparation of catalysts
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrateShow MoreShow Less
PubChem CID 24821
ChEBI CHEBI:86214
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98.0-102.0%
UN Number UN 1477
Solubility Information Very soluble in water,alcohol,and most organic solvents.
IUPAC Name cobalt(2+);dinitrate;hexahydrate
Formula Weight 291.04 (182.94 Anhydrous)
Grade ACS
Odor Odorless
Refractive Index 1.52
Appearance Red crystalline powder
Density 1.87 g/cm3
Chemical Name or Material Cobalt(II) Nitrate Hexahydrate, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY25K009 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number036418.36,036418.A4,036418.22
Lot NumberX31K003 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number036418.36,036418.A4,036418.22
Lot NumberX31K004 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number036418.36,036418.A4,036418.22
Lot NumberQ17K010 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number036418.36,036418.A4,036418.22
Lot NumberN29K031 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number036418.36,036418.A4,036418.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.