missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11453287

Cobalt(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11453287
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11453287

Brand: Thermo Scientific Alfa Aesar A16201.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cobalt(II) sulfate heptahydrate is used in the preparation of pigments, as well as in the manufacture of other cobalt salts, which is used in porcelains and glass. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) sulfate heptahydrate is used in the preparation of pigments, as well as in the manufacture of other cobalt salts, which is used in porcelains and glass. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2oShow MoreShow Less
PubChem CID 61444
IUPAC Name cobalt(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

pH 4
Quantity 500 g
Linear Formula CoSO4·7H2O
UN Number UN3077
Merck Index 14,2448
Solubility Information Soluble in water.
Formula Weight 281.10 (154.99 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 2.03
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249160 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog NumberA16201.0E,A16201.36,A16201.22
Lot Number10241375 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA16201.0E,A16201.36,A16201.22
Lot Number10237381 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16201.0E,A16201.36,A16201.22
Lot Number10163345 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16201.0E,A16201.36,A16201.22
Lot Number10156770 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16201.0E,A16201.36,A16201.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Cobalt(II) sulfate heptahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) sulfate heptahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.