missing translation for 'onlineSavingsMsg'
Learn More
Coenzyme A trilithium salt, 95% Product Code.: 15476899

Coenzyme A trilithium salt, 95%

Product Code. 15476899
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 15476899

Brand: Thermo Scientific Alfa Aesar J65434.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18439-24-2 | C21H33Li3N7O16P3S | 785.329 g/mol

Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis. CoA is involved in the mechanisms of a wide variety of enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coenzyme A facilitates removal of lipid peroxides by increasing mobilization of fatty acids, and promote repair of plasma membranes by activating phospholipid synthesis. CoA is involved in the mechanisms of a wide variety of enzymes
TRUSTED_SUSTAINABILITY

Specifications

CAS 18439-24-2
Color White
Molecular Formula C21H33Li3N7O16P3S
MDL Number MFCD00075848
Quantity 100 mg
Sensitivity Very Hygroscopic
Synonym coenzyme a, trilithium salt
Solubility Information Soluble in water.
InChI Key QSCBPHBAFBVXRK-UHFFFAOYSA-K
SMILES [Li+].[Li+].[Li+].CC(C)(COP(=O)([O-])OP(=O)([O-])OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)[O-])C(C(=O)NCCC(=O)NCCS)O
IUPAC Name trilithium;[5-(6-aminopurin-9-yl)-4-hydroxy-2-[[[[3-hydroxy-2,2-dimethyl-4-oxo-4-[[3-oxo-3-(2-sulfanylethylamino)propyl]amino]butoxy]-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]oxolan-3-yl] hydrogen phosphate
Molecular Weight (g/mol) 785.329
PubChem CID 53442190
Formula Weight 785.33
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Coenzyme A trilithium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ09L018 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberJ65434.MC
Lot NumberZ26K013 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog NumberJ65434.MC
Lot NumberX10K034 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ65434.MC
Lot NumberS26K002 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog NumberJ65434.MC
Lot NumberS07K030 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberJ65434.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Coenzyme A trilithium salt, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.