Learn More
SCAT Collectors for HPLC
Collect several tubes into a single connector - no problem with the right collector! The tube connector is the most flexible connector of the SCAT system.
£44.10 - £88.30
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
10588140
|
SCAT
160139 |
2-in-1 Collector, Sideways | Fittings for 2 x 5.0 - 11.5 mm ID |
£60.65
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10475323
|
SCAT
160134 |
2-in-1 collector, for capillary, 3 x 3.2 mm OD, PTFE/PFA | Fittings for 3 x 3.2 mm OD |
£49.60
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10064784
|
SCAT
160141 |
2-in-1 collector, vertical, PTFE/PFA, 2 x 2.3/3.2 mm OD | Fittings for 2 x 2.3/3.2 mm OD |
£44.10
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10590251
|
SCAT
199010 |
3-in-1 collector for T-piece VWR-Hitachi Chromaster HPLC system | Fittings for capillaries of 2.3/3.2 mm |
£66.20
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10233503
|
SCAT
160130 |
3-in-1 Collector, Sideways | Fittings for 2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID |
£66.20
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
10358729
|
SCAT
160138 |
3-in-1 Collector, Sideways | Fittings for 3 x 5.0 - 11.5 mm ID |
£73.60
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10394123
|
SCAT
160132 |
3-in-1 collector for tube connector, vertical, PTFE/PFA, 3 x 2.3/3.2 mm OD | Fittings for 3 x 2.3/3.2 mm OD |
£49.60
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
15154609
|
SCAT
160128 |
3-in-1 collector for tube connector, vertical, PTFE/PP, 3 x 5.0 - 11.5 mm ID | Fittings for 3 x 5.0 - 11.5 mm ID |
£66.20
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
10723862
|
SCAT
160137 |
8-in-1 collector for tube connector, vertical, PTFE/PFA, 8 x 2.3/3.2 mm OD | Fittings for 8 x 2.3/3.2 mm OD |
£88.30
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10629680
|
SCAT
160129 |
8-in-1 collector for tube connector, vertical, PTFE/PFA/PP, 7 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID | Fittings for 7 x 2,3/3,2 mm OD, 1 x 5,0 - 11,5 mm ID |
£88.30
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||