missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™ VS10-12-1.5
Description
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Recommended for maximising sample volume
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Red | |
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
12 | |
1.5 mm |
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations | |
Comb | |
1 Each | |
50 ÎĽL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1.5mm Thick >
Spot an opportunity for improvement?Share a Content Correction