Learn More
Copper(II) nitrate hemi(pentahydrate), 98%, Thermo Scientific Chemicals
£27.54 - £357.78
Chemical Identifiers
CAS | 19004-19-4 |
---|---|
Molecular Formula | CuN2O6·2.5H2O |
MDL Number | MFCD00149671 |
Synonym | dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427866
|
Thermo Scientific Alfa Aesar
A14004.30 |
250 g |
£27.54
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417866
|
Thermo Scientific Alfa Aesar
A14004.0B |
1000 g |
£77.79
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437866
|
Thermo Scientific Alfa Aesar
A14004.0I |
5000 g |
£357.78
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
Solubility
Soluble in water, ammonia and ethanol
Notes
Incompatible with reducing agents, organic materials and powdered metals.
Chemical Identifiers
19004-19-4 | |
CuN2O6·2.5H2O | |
MFCD00149671 | |
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate |
Specifications
170°C (decomposition) | |
19004-19-4 | |
Cu(NO3)2·2.5H2O | |
UN3085 | |
dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate | |
232.59 (187.55 Anhydrous) | |
Odorless | |
2.32 |
∼115°C | |
CuN2O6·2.5H2O | |
MFCD00149671 | |
14,2643 | |
Soluble in water,ammonia and ethanol | |
98% | |
Hygroscopic | |
Copper(II) nitrate hemi(pentahydrate) |
Safety and Handling
GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 221-838-5
missing translation for 'rtecsNumber' : GL7875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Copper(II) nitrate hemi(pentahydrate)
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P211-Do not spray on an open flame or other ignition source.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Copper(II) nitrate hemi(pentahydrate)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.