Learn More
Corning™ Collagen I, High Concentration, Rat Tail

Sourced from rat tail tendon
Brand: Corning™ 354249
231.80 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Specifications
8 to 11 mg/mL | |
Liquid in 0.02N acetic acid | |
Rat Tail Tendon | |
2°C to 8°C |
Cell Attachment and Proliferation, 3D Gel | |
100 mg | |
Collagen, Rat Tail Type I HC | |
>95% by SDS-PAGE |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only (RUO).