Learn More
Corning™ Costar™ Thermowell™ Plate Holder
GREENER_CHOICE
For Thermo well polycarbonate plates
Brand:  Corning™ 2572
75.43 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10206631
  • £79.40 / Pack of 20
Estimated Shipment: 15-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 96-well egg crate style holder for 1 x 8 Stripwell™ strips
  • Holds twelve 1 x 8 strips
  • Allows each individual well to be positioned back into the plate once broken apart
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate Holder
Holds twelve 1 x 8 strips
Thermo well polycarbonate plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Thermowell™ Plate Holder > Plate Holder for Thermo well polycarbonate plates