Learn More
PSI-6206, MedChemExpress
MedChemExpress PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
Brand: Medchem Express HY-15236
Description
PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.Specifications
PSI-6206 | |
Off-White | |
C10H13FN2O5 | |
RO 2433 GS-331007 | |
O=C(NC1=O)C=CN1[C@H]2[C@](F)([C@@H]([C@H](O2)CO)O)C | |
260.22 | |
Research | |
COVID-19-anti-virus |
863329-66-2 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (384.29 mM; Need ultrasonic) | |
260.22 | |
99.04% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed.