Learn More
Corning™ DeckWorks™ Low Binding Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures
Brand: Corning™ 4137
111.61 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Corning DeckWorks low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.

Specifications
1 to 100 μL | |
Sterile | |
Used for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Yes | |
HDPE |
Filtered | |
Natural | |
Hinged Rack | |
Rack | |
DeckWorks |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
9Hours
8Minutes
3Seconds
34 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
26Hours
8Minutes
3Seconds
34
Your input is important to us. Please complete this form to provide feedback related to the content on this product.