Learn More
Corning™ Magnetic Stir Bar Retriever
GREENER_CHOICE
For Use With: Stirring Models
Brand:  Corning™ 6970SR
10.54 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 10700340
  • £11.60 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
The Corning stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory. The container must be taken off the magnetic stirrer before using the retriever as the high powered magnet in the stirrer cancels out the one in the retriever. The handle is designed for ergonomic and easy retrieval. -The retriever is available one per case.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stirring Models
Accessory, Stir Bar Retriever
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    41
    Hours
    21
    Minutes
    14
    Seconds
    47
  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073
    Expires2025-04-25
    New Offer
    Save Now
Product Content Correction
Product Title
Corning™ Magnetic Stir Bar Retriever > Stir bar retriever