Learn More
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes
GREENER_CHOICE
With white rubber liner
Brand:  Corning™ 99999-18
 View more versions of this product
Product Code. 10602624
  • £419.79 / Pack of 1000
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Made of a phenolic compound. These caps have a white rubber liner.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
18-415
Screw Cap
Phenolic
17.5 mm
1000/Cs.
Culture Tubes
Rubber
Autoclavable
Black
1000/Pk.
Rubber
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes > 18-415