Learn More
Corning™ Costar™ Ultra-Low Attachment Microplates



Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.
Brand: Corning™ 3471
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic

Specifications
2 x 3 | |
Yes | |
Flat | |
Clear | |
Polystyrene | |
Sterile | |
6 | |
Lid | |
1/Pk. | |
24/Cs. |
Clear | |
Polystyrene | |
2.9 mL (Well) | |
9.5 cm2 | |
Rectangular | |
Multi-well Cell Culture Plate | |
Maintaining cells in a suspended, unattached state | |
24/Cs. | |
Individually Wrapped | |
3.48 cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.