missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Ultra-Low Attachment Microplates
GREENER_CHOICEBIOPHARMA

Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.

Brand:  Corning™ 3473

498.82 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10327701

  • £601.00 / Pack of 24
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 x 6
Yes
Flat
Clear
Polystyrene
Sterile
24
Lid
1/Pk.
24/Cs.
Clear
Polystyrene
0.57 mL (Well)
1.9 cm2
Rectangular
Multi-well Cell Culture Plate
Maintaining cells in a suspended, unattached state
24/Cs.
Individually Wrapped
1.56 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number27924016 Certificate TypeCertificate of Quality Date07/04/2025 Catalog Number3473
Lot Number25024028 Certificate TypeCertificate of Quality Date07/04/2025 Catalog Number3473
Lot Number24924036 Certificate TypeCertificate of Quality Date20/03/2025 Catalog Number3473
Lot Number24824016 Certificate TypeCertificate of Quality Date13/02/2025 Catalog Number3473
Lot Number22324002 Certificate TypeCertificate of Quality Date13/02/2025 Catalog Number3473
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Costar™ Ultra-Low Attachment Microplates > 24-well; Ultra-Low Attachment; 1/Pk; 24/Cs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.